Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080554.2 | NP_542121.2 | 6 | 770 | ||
GenBank mRNA | AK005335.1 | - | 6 | 748 | ||
AK011026.1 | - | 6 | 747 | |||
AK029067.1 | - | 5 | 1067 | |||
AK032992.1 | - | 6 | 748 | |||
AK078419.1 | - | 6 | 748 | |||
AK145587.1 | - | 6 | 747 | |||
AK148284.1 | - | 771 | ||||
AK205476.1 | - | 1 | 71 | |||
AK207702.1 | - | 1 | 71 | |||
AK220321.1 | - | 6 | 711 | |||
BC019112.1 | - | 6 | 781 |