Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025920.3 | NP_080196.3 | 2 | 413 | ||
GenBank mRNA | AK086483.1 | - | 2 | 226 | ||
AK140888.1 | - | 356 | ||||
AK167814.1 | - | 2 | 173 | |||
BC063758.1 | - | 2 | 144 | 6334411 | ||
BC066042.1 | - | 2 | 439 | 6413779 |