Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025927.4 | NP_080203.1 | 7 | 852 | ||
GenBank mRNA | AK011163.1 | - | 7 | 721 | ||
AK011605.1 | - | 7 | 711 | |||
AK081922.1 | - | 7 | 713 | |||
AK157771.1 | - | 7 | 714 | |||
AK180833.1 | - | 2 | 186 | |||
AK187738.1 | - | 2 | 187 | |||
BC020130.1 | - | 7 | 697 | 3990098 |