Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081356.3 | NP_001074825.1 | 2 | 245 | ||
NM_001290780.1 | NP_001277709.1 | 2 | 372 | |||
NM_001290781.1 | NP_001277710.1 | 3 | 546 | |||
GenBank mRNA | AA015086.1 | - | 3 | 314 | ||
AJ748652.1 | - | 2 | 193 | |||
AK008848.1 | - | 2 | 203 | |||
AK011632.1 | - | 2 | 211 | |||
AK045401.1 | - | 2 | 203 | |||
AK049296.1 | - | 2 | 183 | |||
AK049488.1 | - | 2 | 216 | |||
AK138582.1 | - | 213 | ||||
AK164163.1 | - | 2 | 200 | |||
BC028317.1 | - | 2 | 183 | |||
BC094639.1 | - | 2 | 199 | 1361961 | ||
CJ062712.1 | - | 2 | 225 | |||
CJ071023.1 | - | 2 | 245 |