Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163687.1 | NP_001157159.1 | 9 | 1043 | ||
NM_025972.4 | NP_080248.2 | 9 | 1049 | |||
GenBank mRNA | AB162194.1 | - | 9 | 1015 | ||
AK008776.1 | - | 9 | 1051 | |||
AK017141.1 | - | 9 | 1026 | |||
AK133658.1 | - | 9 | 1050 | |||
AK146191.1 | - | 9 | 1037 | |||
BC004572.1 | - | 9 | 1019 | 3586255 |