Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291155.1 | NP_001278084.1 | 2 | 327 | ||
NM_025996.5 | NP_080272.1 | 2 | 706 | |||
GenBank mRNA | AK011786.1 | - | 2 | 706 | ||
AK017699.1 | - | 2 | 272 | |||
AK075873.1 | - | 2 | 324 | |||
AK168329.1 | - | 2 | 261 | |||
BB869739.1 | - | 2 | 328 | |||
BC018278.1 | - | 2 | 257 | 4187356 | ||
CX235437.1 | - | 2 | 246 |