Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291155.1 | NP_001278084.1 | 5 | 753 | ||
NM_025996.5 | NP_080272.1 | 5 | 1132 | |||
GenBank mRNA | AK011786.1 | - | 5 | 1132 | ||
AK017699.1 | - | 5 | 698 | |||
AK075873.1 | - | 5 | 750 | |||
AK168329.1 | - | 5 | 687 | |||
BC018278.1 | - | 5 | 683 | 4187356 |