Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011416.2 | NP_035546.2 | 32, 33 | 4819 | ||
NM_026003.2 | NP_080279.1 | 8, 9 | 968 | |||
GenBank mRNA | AK009309.1 | - | 4, 5 | 478 | ||
AK011935.1 | - | 8, 9 | 913 | |||
AK147322.1 | - | 32, 33 | 4968 | |||
BC022975.1 | - | 10, 11 | 1445 | |||
BC056199.1 | - | 22, 23 | 2917 | |||
BC075641.1 | - | 32, 33 | 4801 | 30544092 |