Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026011.3 | NP_080287.1 | 4 | 537 | ||
GenBank mRNA | AB185206.1 | - | 4 | 341 | ||
AK005999.1 | - | 4 | 384 | |||
AK009279.1 | - | 4 | 474 | |||
AK012024.1 | - | 4 | 501 | |||
AK030689.1 | - | 4 | 517 | |||
AK075941.1 | - | 4 | 498 | |||
AK144177.1 | - | 4 | 508 | |||
AK146307.1 | - | 4 | 503 | |||
AK149822.1 | - | 4 | 487 | |||
AK151036.1 | - | 4 | 498 | |||
AK152798.1 | - | 4 | 515 | |||
AK159311.1 | - | 3 | 184 | |||
AK159348.1 | - | 4 | 503 | |||
AK169929.1 | - | 4 | 471 | |||
AK170645.1 | - | 4 | 503 | |||
AK201340.1 | - | 2 | 92 | |||
AK206384.1 | - | 2 | 92 | |||
AK219184.1 | - | 2 | 92 | |||
AK219830.1 | - | 2 | 92 | |||
BC013719.1 | - | 4 | 486 | 3983809 | ||
CT010337.1 | - | 4 | 341 |