Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 5 | 870 | ||
GenBank mRNA | AB086655.1 | - | 5 | 849 | ||
AF477481.1 | - | 5 | 870 | |||
AK012041.1 | - | 5 | 847 | |||
AK017609.1 | - | 5 | 844 | |||
AK028287.1 | - | 5 | 842 | |||
AK049163.1 | - | 5 | 842 | |||
AK076527.1 | - | 4 | 952 | |||
AK083015.1 | - | 5 | 845 | |||
AK208010.1 | - | 1 | 43 | |||
BC013842.1 | - | 5 | 825 | |||
BC024485.1 | - | 2 | 121 | |||
BC048076.1 | - | 5 | 870 | 6307476 |