Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026025.2 | NP_080301.1 | 2, 3 | 207 | ||
GenBank mRNA | AB095989.1 | - | 2, 3 | 240 | ||
AK012589.1 | - | 2, 3 | 232 | |||
AK012852.1 | - | 2, 3 | 208 | |||
AK042283.1 | - | 2, 3 | 191 | |||
AK170136.1 | - | 2, 3 | 219 | |||
BC058368.1 | - | 2, 3 | 241 | 6819507 |