Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144517.4 | NP_653100.1 | 14 | 1159 | ||
GenBank mRNA | AK012836.1 | - | 14 | 1110 | ||
AK076128.1 | - | 14 | 1107 | |||
AK083084.1 | - | 14 | 1130 | |||
BC020138.1 | - | 14 | 1118 | 3983701 |