Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 11 | 1206 | ||
GenBank mRNA | AK013554.1 | - | 11 | 1158 | ||
AK021316.1 | - | 4 | 313 | |||
AK028937.1 | - | 11 | 1132 | |||
AK028945.1 | - | 11 | 1109 | |||
AK030280.1 | - | 11 | 1146 | |||
AK076409.1 | - | 2 | 140 | |||
AK134887.1 | - | 10 | 1865 | |||
AK210387.1 | - | 2 | 160 | |||
AY029531.1 | - | 11 | 1107 | |||
BC003946.1 | - | 11 | 1093 | 3487343 |