Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026061.2 | NP_080337.1 | 4 | 339 | ||
GenBank mRNA | AK010177.1 | - | 4 | 329 | ||
AK013516.1 | - | 4 | 339 | |||
AK028070.1 | - | 4 | 350 | |||
AK212374.1 | - | 2 | 49 | |||
BC043019.1 | - | 4 | 339 | 5684256 |