Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026064.2 | NP_080340.2 | 4 | 1160 | ||
GenBank mRNA | AK013776.1 | - | 4 | 847 | ||
AK028020.1 | - | 4 | 796 | |||
AK045160.1 | - | 4 | 813 | |||
AK151265.1 | - | 4 | 839 | |||
AK159151.1 | - | 4 | 819 | |||
AK167553.1 | - | 4 | 831 | |||
AK168016.1 | - | 4 | 813 | |||
BC006719.1 | - | 2 | 526 | |||
BC010265.1 | - | 3 | 745 | 3588394 | ||
BC020282.1 | - | 4 | 823 | |||
BC029180.1 | - | 4 | 795 | 2599135 | ||
BC089050.1 | - | 3 | 1217 |