Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 46 | 6100 | ||
NM_026082.5 | NP_080358.3 | 45 | 6010 | |||
GenBank mRNA | AK013336.1 | - | 4 | 389 | ||
AK014226.1 | - | 2 | 213 | |||
AK031531.1 | - | 1 | 3627 | |||
AK122554.2 | - | 15 | 1744 | |||
AK150619.1 | - | 13 | 1691 | |||
AK152431.1 | - | 13 | 1691 | |||
BC024823.1 | - | 16 | 2112 | |||
BC024917.1 | - | 10 | 1265 | |||
BC055032.1 | - | 5 | 506 | |||
BC086672.1 | - | 26 | 3518 | 30647593 | ||
BC094420.1 | - | 34 | 4301 |