Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026109.2 | NP_080385.2 | 4 | 495 | ||
GenBank mRNA | AK007381.1 | - | 4 | 504 | ||
AK019032.1 | - | 4 | 484 | |||
BC132664.1 | - | 4 | 467 | 40130941 | ||
BC132666.1 | - | 4 | 467 | 40130943 | ||
BY707479.1 | - | 500 |