Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410127L17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026120.4 | NP_080396.2 | 2 | 546 | ||
GenBank mRNA | AI509964.1 | - | 2 | 332 | ||
AK010784.1 | - | 2 | 431 | |||
BB656580.1 | - | 2 | 562 | |||
BC019559.1 | - | 2 | 736 | |||
BC043335.1 | - | 2 | 500 | 5030742 | ||
BC092281.1 | - | 2 | 378 | 4950426 |