Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000898.4 | NP_000889.3 | 9 | 1157 | ||
XM_005272608.3 | XP_005272665.1 | 11 | 1257 | |||
XM_017029523.1 | XP_016885012.1 | 11 | 1307 | |||
XM_017029524.1 | XP_016885013.1 | 10 | 1117 | |||
GenBank mRNA | AK298942.1 | - | 9 | 1094 | ||
AK312679.1 | - | 9 | 1132 | |||
BC022494.1 | - | 9 | 1103 | 4800208 | ||
M69177.1 | - | 9 | 1058 | |||
S62734.1 | - | 9 | 981 |