Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285831.1 | NP_001272760.1 | 6 | 746 | ||
NM_027379.3 | NP_081655.2 | 6 | 746 | |||
GenBank mRNA | AK005531.1 | - | 6 | 720 | ||
AK011187.1 | - | 6 | 721 | |||
AK014405.1 | - | 6 | 719 | |||
AK014486.1 | - | 6 | 719 | |||
AK030067.1 | - | 6 | 677 | |||
AK033674.1 | - | 6 | 689 | |||
AK039083.1 | - | 2 | 164 | |||
BC007178.1 | - | 6 | 694 | 3495305 |