Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026170.3 | NP_080446.1 | 5 | 366 | ||
GenBank mRNA | AB093571.1 | - | 5 | 265 | ||
AK003239.1 | - | 5 | 353 | |||
AK004629.1 | - | 5 | 320 | |||
AK040727.1 | - | 2 | 518 | |||
AK084385.1 | - | 5 | 313 | |||
AK088157.1 | - | 5 | 321 | |||
AK151239.1 | - | 5 | 320 | |||
AK173123.1 | - | 5 | 355 | |||
BC005516.1 | - | 5 | 366 | 3494155 | ||
BC083144.1 | - | 5 | 286 | 30432732 | ||
CT010230.1 | - | 5 | 265 |