Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026201.3 | NP_080477.1 | 7, 8 | 834 | ||
GenBank mRNA | AF465615.1 | - | 7, 8 | 725 | ||
AK012111.1 | - | 7, 8 | 725 | |||
AK215105.1 | - | 2, 3 | 267 | |||
BC010199.1 | - | 7, 8 | 712 | |||
BC034174.1 | - | 7, 8 | 735 | |||
BC051052.1 | - | 7, 8 | 736 | |||
BC060130.1 | - | 7, 8 | 722 | |||
BC079652.1 | - | 7, 8 | 708 | 6844310 |