Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026210.4 | NP_080486.1 | 3, 4 | 342 | ||
GenBank mRNA | AK007707.1 | - | 3, 4 | 258 | ||
AK017572.1 | - | 3, 4 | 257 | |||
AK050027.1 | - | 3, 4 | 378 | |||
AK077277.1 | - | 3, 4 | 344 | |||
AK150936.1 | - | 3, 4 | 334 | |||
BC115504.1 | - | 3, 4 | 322 | 40101013 | ||
BC115505.1 | - | 3, 4 | 322 | 40101015 |