Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026220.4 | NP_080496.1 | 7 | 1924 | ||
GenBank mRNA | AK014483.1 | - | 7 | 1917 | ||
AK032586.1 | - | 7 | 1925 | |||
AK050561.1 | - | 7 | 1925 | |||
AK088766.1 | - | 7 | 1922 | |||
AK161000.1 | - | 7 | 1925 | |||
AK168801.1 | - | 7 | 1930 |
Gene Symbol: Mfap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003440.1 | - | 433 | |||
AK169029.1 | - | 1928 |