Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026232.3 | NP_080508.1 | 3 | 261 | ||
GenBank mRNA | AK015679.1 | - | 3 | 262 | ||
AK017035.1 | - | 3 | 262 | |||
AK030950.1 | - | 3 | 230 | |||
AK090086.1 | - | 3 | 199 | |||
AK151334.1 | - | 3 | 230 | |||
AK151420.1 | - | 3 | 198 | |||
AK159122.1 | - | 3 | 259 | |||
AK159732.1 | - | 3 | 259 | |||
BC022676.1 | - | 3 | 201 | 4238839 |