Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163316.1 | NP_001156788.1 | 4 | 303 | ||
NM_026269.2 | NP_080545.2 | 4 | 494 | |||
GenBank mRNA | AK003551.1 | - | 4 | 443 | ||
BC019557.1 | - | 4 | 285 | 4163641 | ||
CJ048159.1 | - | 4 | 302 |