Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026276.3 | NP_080552.3 | 4 | 694 | ||
GenBank mRNA | AK007737.1 | - | 4 | 695 | ||
AK008554.1 | - | 3 | 497 | |||
AK010557.1 | - | 4 | 654 | |||
AK013111.1 | - | 4 | 648 | |||
AK014188.1 | - | 4 | 637 | |||
AK018191.1 | - | 3 | 340 | |||
AK028700.1 | - | 4 | 651 | |||
AK029523.1 | - | 4 | 636 | |||
AK031766.1 | - | 3 | 516 | |||
AK076099.1 | - | 4 | 649 | |||
AK076866.1 | - | 4 | 668 | |||
AK081818.1 | - | 4 | 671 | |||
AK088977.1 | - | 3 | 512 | |||
AK144671.1 | - | 4 | 668 | |||
BC030043.1 | - | 4 | 629 | 1348651 | ||
BC038013.1 | - | 4 | 665 | 3497754 | ||
BC049851.1 | - | 4 | 655 | 3481972 |