Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8549
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
6
932
NM_001204137.1
NP_001191066.1
6
932
NM_001204138.1
NP_001191067.1
6
932
NM_001204139.1
NP_001191068.1
6
932
NM_001204140.1
NP_001191069.1
6
932
NM_001204141.1
NP_001191070.1
6
932
NM_001204142.1
NP_001191071.1
6
932
NM_001204143.1
NP_001191072.1
6
713
NM_001204151.2
NP_001191080.1
6
932
NM_001323942.1
NP_001310871.1
6
932
NM_001323947.1
NP_001310876.1
6
932
NM_001323949.1
NP_001310878.1
6
924
NM_001323950.1
NP_001310879.1
6
932
NM_001323951.1
NP_001310880.1
6
932
NM_001323952.1
NP_001310881.1
6
932
NM_001323953.1
NP_001310882.1
6
939
NM_001323954.1
NP_001310883.1
6
932
NM_002384.2
NP_002375.1
6
932
NM_015844.2
NP_056669.2
6
932
NM_015845.3
NP_056670.2
6
932
NM_015846.3
NP_056671.2
6
932
NM_015847.3
NP_056723.2
6
932
XM_005258271.2
XP_005258328.1
6
679
XM_006722456.2
XP_006722519.1
6
679
XM_011525991.1
XP_011524293.1
6
679
XM_011525993.2
XP_011524295.1
6
679
XM_011525994.2
XP_011524296.1
6
679
XM_011525998.2
XP_011524300.1
6
679
XM_011525999.1
XP_011524301.1
6
679
XM_011526001.1
XP_011524303.1
6
679
XM_011526002.1
XP_011524304.1
6
679
XM_011526003.2
XP_011524305.1
6
679
XM_011526006.1
XP_011524308.1
6
679
XM_011526007.1
XP_011524309.1
6
679
XM_017025751.1
XP_016881240.1
6
679
XM_017025752.1
XP_016881241.1
6
679
XM_017025753.1
XP_016881242.1
6
679
XM_017025754.1
XP_016881243.1
6
679
XM_017025755.1
XP_016881244.1
6
679
XM_017025756.1
XP_016881245.1
6
679
XM_017025757.1
XP_016881246.1
6
679
XM_017025758.1
XP_016881247.1
6
679
XM_017025759.1
XP_016881248.1
6
679
XM_017025760.1
XP_016881249.1
6
679
XM_017025761.1
XP_016881250.1
6
679
XM_017025762.1
XP_016881251.1
6
679
XM_017025763.1
XP_016881252.1
6
679
XM_017025764.1
XP_016881253.1
6
679
XM_017025765.1
XP_016881254.1
6
679
XM_017025766.1
XP_016881255.1
6
679
XM_017025767.1
XP_016881256.1
6
679
XM_017025768.1
XP_016881257.1
6
679
XM_017025769.1
XP_016881258.1
6
679
XM_017025770.1
XP_016881259.1
6
679
XM_017025771.1
XP_016881260.1
6
679
XM_017025772.1
XP_016881261.1
6
679
XM_017025773.1
XP_016881262.1
6
679
XM_017025774.1
XP_016881263.1
6
679
XM_017025775.1
XP_016881264.1
6
679
XM_017025776.1
XP_016881265.1
6
679
XM_017025777.1
XP_016881266.1
6
679
GenBank mRNA
AB209765.1
-
3
269
AF078830.1
-
5
495
AF078831.1
-
5
495
AF078832.1
-
5
495
AF078833.1
-
5
495
AJ564845.1
-
6
634
AK225889.1
-
6
842
AK225948.1
-
6
638
AK291519.1
-
6
933
AK295401.1
-
6
664
AK300759.1
-
6
713
AK302004.1
-
6
624
BC033242.1
-
6
658
5432943
BC047353.1
-
6
617
Y10746.1
-
6
634
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0