Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026344.3 | NP_080620.1 | 4 | 1056 | ||
GenBank mRNA | AK011199.1 | - | 4 | 1074 | ||
AK018638.1 | - | 4 | 1057 | |||
AK051026.1 | - | 4 | 1076 | |||
AK080747.1 | - | 3 | 1267 | |||
BC014746.1 | - | 3 | 952 | |||
BC031124.1 | - | 4 | 1043 | 5029876 |