Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026366.2 | NP_080642.1 | 5 | 575 | ||
GenBank mRNA | AK012019.1 | - | 5 | 515 | ||
AK145617.1 | - | 5 | 527 | |||
AK160298.1 | - | 5 | 538 | |||
AY456393.1 | - | 5 | 575 | |||
BC116393.1 | - | 5 | 545 | 40101877 | ||
BC116394.1 | - | 5 | 545 | 40101881 |