Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026374.3 | NP_080650.1 | 5, 6 | 325 | ||
GenBank mRNA | AK012364.1 | - | 5, 6 | 312 | ||
AK013858.1 | - | 5, 6 | 371 | |||
AK031892.1 | - | 5, 6 | 380 | |||
AK044885.1 | - | 5, 6 | 371 | |||
AK078003.1 | - | 5, 6 | 312 | |||
AK135449.1 | - | 5, 6 | 400 | |||
AK135539.1 | - | 5, 6 | 312 | |||
AK157129.1 | - | 5, 6 | 380 | |||
BC024718.1 | - | 5, 6 | 325 | 5012365 |