Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026402.3 | NP_080678.1 | 5 | 551 | ||
GenBank mRNA | AB079386.1 | - | 5 | 280 | ||
AK005266.1 | - | 5 | 558 | |||
AK011409.1 | - | 5 | 552 | |||
AK078877.1 | - | 5 | 544 | |||
AK150914.1 | - | 5 | 552 | |||
AK159231.1 | - | 5 | 552 | |||
AK220135.1 | - | 1 | 31 | |||
BC010809.1 | - | 5 | 549 | 4241199 | ||
BC030053.1 | - | 3 | 1273 |