Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026422.2 | NP_080698.1 | 6 | 1027 | ||
GenBank mRNA | AK010018.1 | - | 6 | 758 | ||
BC019787.1 | - | 6 | 771 | 4167280 |