Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099855.1 | NP_001093325.1 | 16 | 1870 | ||
NM_001171876.1 | NP_001165347.1 | 17 | 1918 | |||
NM_001171877.1 | NP_001165348.1 | 12 | 1735 | |||
NM_001171878.1 | NP_001165349.1 | 13 | 1852 | |||
NM_001171879.1 | NP_001165350.1 | 14 | 1900 | |||
NM_005369.4 | NP_005360.3 | 13 | 1852 | |||
XM_005262413.4 | XP_005262470.1 | 18 | 2066 | |||
XM_011531339.2 | XP_011529641.1 | 18 | 2125 | |||
XM_017029529.1 | XP_016885018.1 | 17 | 1887 | |||
XM_017029530.1 | XP_016885019.1 | 17 | 1887 | |||
XM_017029531.1 | XP_016885020.1 | 17 | 1820 | |||
XM_017029532.1 | XP_016885021.1 | 14 | 1977 | |||
GenBank mRNA | AB085901.1 | - | 13 | 1529 | ||
AB085902.1 | - | 14 | 1577 | |||
AK296488.1 | - | 12 | 1735 | |||
AK296529.1 | - | 16 | 1970 | |||
AK302957.1 | - | 18 | 1866 | |||
AK313900.1 | - | 13 | 1712 | |||
AK316416.1 | - | 17 | 1906 | |||
AL117234.1 | - | 15 | 1753 | |||
J03639.1 | - | 585 | ||||
X12556.1 | - | 13 | 1703 | |||
X13230.1 | - | 457 |