Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026438.4 | NP_080714.2 | 5 | 475 | ||
GenBank mRNA | AK008575.1 | - | 5 | 476 | ||
AK020291.1 | - | 5 | 479 | |||
AK088397.1 | - | 5 | 471 | |||
AK145640.1 | - | 5 | 525 | |||
AK146300.1 | - | 5 | 438 | |||
AK151512.1 | - | 5 | 466 | |||
AK219020.1 | - | 2 | 134 | |||
BC010468.1 | - | 5 | 409 | 3585780 | ||
CT010203.1 | - | 5 | 368 |