Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026442.3 | NP_080718.1 | 4 | 409 | ||
GenBank mRNA | AK002667.1 | - | 4 | 306 | ||
AK020626.1 | - | 4 | 293 | |||
AK168728.1 | - | 4 | 356 | |||
BC048547.1 | - | 4 | 390 | |||
BC056653.1 | - | 4 | 283 | 5068125 |