Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026443.4 | NP_080719.2 | 4 | 1354 | ||
GenBank mRNA | AK006148.1 | - | 4 | 1275 | ||
AK010971.1 | - | 4 | 1311 | |||
AK012058.1 | - | 4 | 1321 | |||
AK078381.1 | - | 4 | 1355 | |||
AK167990.1 | - | 4 | 1223 | |||
AK216258.1 | - | 1 | 324 | |||
BC012242.1 | - | 1 | 768 | |||
BC019205.1 | - | 4 | 1364 | |||
BC058737.1 | - | 4 | 1304 | 5400435 |