Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026464.2 | NP_080740.2 | 7 | 1043 | ||
GenBank mRNA | AK017695.1 | - | 7 | 1017 | ||
AK018469.1 | - | 7 | 1024 | |||
AK075958.1 | - | 7 | 1043 | |||
AK146093.1 | - | 7 | 1062 | |||
BC011484.1 | - | 5 | 709 | |||
BC064750.1 | - | 7 | 1002 | |||
BC131932.1 | - | 7 | 1037 | 40130209 | ||
BC131934.1 | - | 7 | 1037 | 40130211 |