Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023402.2 | NP_075891.1 | 4 | 813 | ||
GenBank mRNA | AK002885.1 | - | 4 | 815 | ||
AK005133.1 | - | 4 | 855 | |||
AK146115.1 | - | 4 | 831 | |||
AK160123.1 | - | 4 | 856 | |||
AK168316.1 | - | 4 | 814 | |||
BC028878.1 | - | 4 | 795 | 4218748 | ||
BC099425.1 | - | 3 | 784 | 30847912 |