Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175127.2 | NP_780336.1 | 3 | 464 | ||
GenBank mRNA | AK076211.1 | - | 3 | 470 | ||
AK077653.1 | - | 3 | 477 | |||
AK122295.1 | - | 3 | 449 | |||
AK162091.1 | - | 3 | 433 | |||
AK162188.1 | - | 3 | 345 | |||
BC055458.1 | - | 2 | 166 | |||
BC094542.1 | - | 3 | 464 | 6408744 |