Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024214.2 | NP_077176.1 | 2, 3 | 370 | ||
GenBank mRNA | AK002902.1 | - | 2, 3 | 291 | ||
AK160168.1 | - | 2, 3 | 273 | |||
AK181461.1 | - | 2, 3 | 89 | |||
AK185977.1 | - | 89 | ||||
BC002087.1 | - | 2, 3 | 279 | 3484983 | ||
BC099449.1 | - | 2, 3 | 277 | 30848526 | ||
DN174518.1 | - | 2, 3 | 370 |