Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164600.1 | NP_001158072.1 | 11 | 1400 | ||
NM_028364.2 | NP_082640.2 | 12 | 1481 | |||
NM_133691.5 | NP_598452.2 | 11 | 1430 | |||
GenBank mRNA | AK012434.1 | - | 11 | 1368 | ||
AK075960.1 | - | 11 | 1398 | |||
AK146534.1 | - | 12 | 1452 | |||
AK149632.1 | - | 12 | 1447 | |||
AK151526.1 | - | 11 | 1394 | |||
AK168789.1 | - | 11 | 1397 | |||
AK178088.1 | - | 2 | 168 | |||
AK182094.1 | - | 2 | 168 | |||
AK207327.1 | - | 2 | 168 | |||
BC010601.1 | - | 11 | 1400 | 3257153 |