Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133692.2 | NP_598453.1 | 11 | 1223 | ||
GenBank mRNA | AF294329.1 | - | 11 | 1237 | ||
AK010805.1 | - | 11 | 1218 | |||
AK012386.1 | - | 3 | 209 | |||
AK031778.1 | - | 11 | 1225 | |||
AK088385.1 | - | 11 | 1216 | |||
AK162240.1 | - | 6 | 552 | |||
AK168179.1 | - | 11 | 1211 | |||
BC055325.1 | - | 11 | 1226 | 6402059 |