Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198295.2 | NP_938037.2 | 6 | 513 | ||
GenBank mRNA | AK035946.1 | - | 6 | 461 | ||
AK042787.1 | - | 6 | 456 | |||
AK048502.1 | - | 6 | 469 | |||
AK050932.1 | - | 6 | 396 | |||
AK129451.1 | - | 6 | 407 | |||
AK140709.1 | - | 6 | 514 | |||
AK157865.1 | - | 6 | 464 |