Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026519.3 | NP_080795.1 | 4 | 527 | ||
GenBank mRNA | AK003668.1 | - | 4 | 495 | ||
AK012047.1 | - | 4 | 528 | |||
AK078165.1 | - | 4 | 528 | |||
AK167745.1 | - | 4 | 479 | |||
BC051926.1 | - | 4 | 483 | 5710720 |