Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026549.3 | NP_080825.1 | 6 | 957 | ||
GenBank mRNA | AK003339.1 | - | 6 | 966 | ||
AK077948.1 | - | 6 | 958 | |||
AK151495.1 | - | 6 | 961 | |||
AK154811.1 | - | 6 | 973 | |||
BC025090.1 | - | 3 | 421 | |||
BC038524.1 | - | 3 | 427 | |||
BC115797.1 | - | 6 | 956 | 40103840 |