Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam92a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026558.4 | NP_080834.3 | 4 | 676 | ||
GenBank mRNA | AK013129.1 | - | 4 | 517 | ||
AK019574.1 | - | 4 | 504 | |||
AK032690.1 | - | 4 | 658 | |||
AK078438.1 | - | 4 | 657 | |||
AK088705.1 | - | 4 | 657 | |||
BC020162.1 | - | 470 | 3991990 | |||
BC055848.1 | - | 4 | 1512 |