Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085407.1 | NP_001078876.1 | 3 | 342 | ||
NM_001085408.1 | NP_001078877.1 | 2 | 273 | |||
NM_026563.3 | NP_080839.2 | 4 | 492 | |||
GenBank mRNA | AK014885.1 | - | 4 | 461 | ||
AK021273.1 | - | 4 | 483 | |||
AK082787.1 | - | 3 | 313 | |||
AK163690.1 | - | 2 | 244 | |||
AW743523.1 | - | 2 | 223 | |||
BC005612.1 | - | 2 | 264 | |||
BC031199.1 | - | 2 | 222 | 5064948 | ||
BC061037.1 | - | 3 | 309 |