Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145337.2 | NP_001138809.1 | 3, 4 | 228 | ||
NM_001145339.2 | NP_001138811.1 | 3, 4 | 478 | |||
NM_002392.5 | NP_002383.2 | 3, 4 | 478 | |||
XM_005268872.4 | XP_005268929.1 | 3, 4 | 406 | |||
GenBank mRNA | AF092843.1 | - | 3, 4 | 186 | ||
AF092844.1 | - | 3, 4 | 186 | |||
AF385323.1 | - | 2, 3 | 154 | |||
AF385324.1 | - | 2, 3 | 154 | |||
AF385326.1 | - | 2, 3 | 154 | |||
AJ430612.1 | - | 2, 3 | 154 | |||
AJ430614.1 | - | 2, 3 | 154 | |||
AJ491698.1 | - | 2, 3 | 154 | |||
AJ491700.1 | - | 2, 3 | 154 | |||
AJ550518.1 | - | 2, 3 | 154 | |||
AJ550519.1 | - | 2, 3 | 154 | |||
AK290341.1 | - | 3, 4 | 428 | |||
AM902646.1 | - | 2, 3 | 154 | |||
AM902650.1 | - | 2, 3 | 154 | |||
AM902651.1 | - | 2, 3 | 154 | |||
AM902654.1 | - | 2, 3 | 154 | |||
AM902655.1 | - | 2, 3 | 154 | |||
AM902659.1 | - | 2, 3 | 154 | |||
AM902660.1 | - | 2, 3 | 154 | |||
AM902666.1 | - | 2, 3 | 154 | |||
AM902667.1 | - | 2, 3 | 154 | |||
AM902668.1 | - | 2, 3 | 154 | |||
AM902669.1 | - | 2, 3 | 154 | |||
AM902672.1 | - | 2, 3 | 154 | |||
AM902678.1 | - | 2, 3 | 154 | |||
AM902680.1 | - | 2, 3 | 154 | |||
AM902681.1 | - | 2, 3 | 154 | |||
BC152384.1 | - | 2, 3 | 157 | |||
BC152390.1 | - | 2, 3 | 157 | |||
BE296905.1 | - | 3, 4 | 443 | |||
BT007258.1 | - | 2, 3 | 154 | |||
EU076747.1 | - | 3, 4 | 228 | |||
EU076749.1 | - | 3, 4 | 228 | |||
HM213901.1 | - | 2, 3 | 154 | |||
HY174841.1 | - | 480 | ||||
JX015379.1 | - | 2, 3 | 154 | |||
JX015380.1 | - | 2, 3 | 154 | |||
JX015387.1 | - | 2, 3 | 154 | |||
JX015389.1 | - | 2, 3 | 154 | |||
JX015390.1 | - | 2, 3 | 154 | |||
JX015392.1 | - | 2, 3 | 154 | |||
M92424.1 | - | 3, 4 | 465 | |||
U33203.1 | - | 2, 3 | 154 | |||
Z12020.1 | - | 3, 4 | 465 |