Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026601.3 | NP_080877.3 | 5 | 611 | ||
GenBank mRNA | AK004328.1 | - | 4 | 199 | ||
AK020099.1 | - | 5 | 485 | |||
BC024619.1 | - | 5 | 599 | 4923664 | ||
BC100401.1 | - | 5 | 575 | |||
CB952524.1 | - | 3 | 116 | |||
CK620463.1 | - | 5 | 499 |